The XlinkX software supports exporting results to xiView, which is a web-based crosslink visualization tool and requires several files for full functionality as in the following figure.
Procedure
- Open the results, and select the checkboxes for all crosslinkers, peptides, proteins, and MS/MS Spectra associated with crosslink spectrum matches to export.
- Select File > Export > To mzIdentML.
- In the Export to mzIdentML dialog box, select Export.
- Export all checked spectra to mzML format.
- Generate FASTA and CVS files with xiNET export.
- Upload all files to xiView.
For information about the software, refer to https://xiview.org/xiNET_website/index.php.
For information about mzIdentML format, refer to http://www.psidev.info/mzidentml#mzid12.