This procedure is related to compensation voltage, which is part of FAIMS™ support.
Procedure
- Open the results file.
- Select the Report Item Distribution icon, and then select the Histogram tab.
- For Data Source, select CSMs - Comp Voltage.
This procedure is related to compensation voltage, which is part of FAIMS™ support.