The XlinkX software supports showing decoy hits in the histogram view.
Procedure
- To show the decoy hits, do the following:
- Open the results file.
- Select the Report Item Distribution icon, and then select the Histograms tab.
- For Data Source, select CSMs - XlinkX Score.
- Under Legend Options, for Show Legend, select Bottom.
- Under Series Options, for Show Decoy Items, select True.
- To restrict display by a specific XlinkX score cutoff, do the following:
- Open the results file.
- Select the Report Item Distribution icon, and then select the Histograms tab.
- For Data Source, select CSMs - D XlinkX Score.