The XlinkX software includes a Decoy CSMs (crosslink spectrum matches) table.
Procedure
- Open the results file.
- Select the Visible Tables icon.
- In the Visible Tables dialog, select Decoy CSMs, and then select OK.
The XlinkX software includes a Decoy CSMs (crosslink spectrum matches) table.