The XlinkX/PD Consensus Validator node to set the FDR thresholds.
Parameter | Definition |
|---|---|
Crosslink FDR threshold | Maximum FDR required for a crosslink to pass. Minimum value = 0; Maximum value = 1 |
CSM FDR threshold | Maximum FDR rate required for a crosslink spectrum match to pass. Minimum value = 0; Maximum value = 1 |