The XlinkX/PD Crosslink Export node exports the list of crosslinked peptides to a xiNET file, which you can import into the xiNET viewer, a web-based crosslink viewer1).
Parameter | Definition |
|---|---|
xiNET Output Files Directory | Opens the Browse for Folder dialog box so that you can browse to the location where the xiNET files are saved. |
Only Master Proteins | Determines whether the XlinkX algorithm exports only those crosslinked peptides from master proteins or crosslinked peptides from all proteins to the xiNET file.
|