The XlinkX/PD Crosslink Grouping node lists the unique crosslinked peptides.
Parameter | Definition |
|---|---|
Ignore reporter scan identification | This option is active for the MS2_MS2 and MS2_MS3 workflows; when set to true, upon finalizing the peptide pair, the software will not take any identification from the reporter scan into account. Default: false |