The XlinkX/PD Validator node determines the FDR for the crosslinks.
Parameter | Description |
|---|---|
FDR Threshold | Specifies an FDR threshold value that determines whether a crosslinked peptide pair passes. The application reports crosslinks with an FDR that is equal to or lower than this value. Range: 0–1; default: 0.01 |