The consensus workflow contains the following two extra nodes:
- XlinkX/PD Crosslink Grouping node—which lists the unique crosslinked peptides
- XlinkX/PD Consensus Validator node—to set the FDR thresholds
For information on the available consensus nodes, see Consensus workflow nodes, and refer to the Proteome Discoverer Help.