A monolink is a crosslinker that only attaches to one peptide. If you do not see a monolink that you are interested in on the Chemical Modifications view, you can create your own. Define a monolink in the Chemical Modifications view as you would an ordinary modification.
For information, refer to the section “Defining Chemical Modifications,” in the Proteome Discoverer User Guide.