Use the following procedure to add cleavable crosslinkers.
Procedure
- To open the Chemical Modifications view, do one of the following:
- Select Administration > Maintain Chemical Modifications.
- Select Maintain Chemical Modifications
.
- Define the modification (refer to the topics on “Defining Chemical Modifications,” in “Configuring Proteome Discoverer” in the Proteome Discoverer User Guide).
- Ensure that the chemical formula in the Substitution field is correct. The XlinkX algorithm uses only the formula to calculate the mass of the modification.
- Ensure that the target amino acid of the crosslinker is set.
- The following image shows the addition of a new crosslinker called DSSO_new in the Chemical Modifications view.
- Select Add to add the crosslinker fragments as necessary for cleavable crosslinkers and to enable the chemical modification in the XlinkX/PD Detect node.
- The Add Chemical Modification dialog box opens to the General page.
- On the General page, do the following
- For Name, enter the name of the crosslinker.
- For Abbreviation, enter the abbreviation of the name of the crosslinker.
- For Delta Mass [Da] (the monoisotopic mass), enter a value.
- For Delta Average Mass [Da], enter a value.
- For Substitution, enter the chemical formula of the crosslinker.
- Select the targeted amino acid.
- Select the Crosslinking tab and do the following:
- Select Cleavable Crosslink from the dropdown list.
- The application automatically calculates the fragment masses. If it is not possible to calculate a mass, it displays an error message and does not add the fragment to the list.
- (Optional) To change the information for a fragment, double-click the field, and then enter the new text.
- (Optional) To remove a fragment, select Delete
on the row.
- After adding all the fragments, you can connect the fragments.
For the identification, you must have knowledge of the possible fragment combinations in a cleaved crosslink. - Select a fragment, and then select Add Selected Fragment.
- The first selected fragment is displayed under Left Fragment.
- Select another fragment, and then select Add Selected Fragment.
- The second selected fragment is displayed under Right Fragment.
- Connect any additional fragments if needed.
- Select OK.