The Shutting Motor Test checks the mechanical stability of the shutter motor and filter paddle.
Follow these steps
Install the diagnostic cell in the detector.
Turn on the detector lamps (one or both).
Run the test from the Chromeleon software. Refer to the Chromeleon Help.
Depends on the test result:
Test result | Action |
---|---|
Passed | Reinstall the flow cell of your application. |
Failed | Contact Thermo Fisher Scientific Technical Support. |