Allowed concentrations (standard system configuration):
System | Chloride | Buffer | Remarks |
---|---|---|---|
Vanquish Core | 0.1 mol/L or less | 1 mol/L or less | High chloride concentration: The application time should be as short as possible. Flush the system thoroughly after these applications. |
Vanquish Horizon Vanquish Flex | 1 mol/L or less | - |