Allowed pH ranges (standard system configuration):
System
(Standard Configuration)
Allowed pH ranges
Remarks
Vanquish Core
1-13
  • pH value of 2 (Vanquish Horizon/Flex): Short-term use only. The application time should be as short as possible. Flush the system thoroughly after these applications.
  • pH value of 1-2 (Vanquish Core): The application time should be as short as possible. Flush the system thoroughly after these applications.
  • pH values higher than 9.5 with optical detectors: Avoid using mobile phases with a pH value higher than 9.5 together with optical detectors. This can impair the functionality and optical performance of the detector flow cell.
Vanquish Horizon
Vanquish Flex
2-12