The Chromeleon software supports diagnostic tests for the detector. You can use these tests to check and monitor the performance and reliability of certain components.
Depending on the Vanquish system configuration and Chromeleon version, the available tests may vary. If a test is grayed out in the Chromeleon software, the test is currently not available.
When a test is running, the LED bar on the module shows a blue running light.