Prerequisites
The lamp house cover must be installed.
TIP
If you remove the lamp house cover while the lamp is turned on, the lamp is turned off automatically.
When using both lamps, it is recommended to turn on the UV lamp prior to the VIS lamp.
Follow these steps
  1. Press the VIS button on the keypad of the detector.
    - or –
    In the chromatography data system, turn on the lamp on the ePanel for the detector.
  2. Allow the lamp and optics to warm up and stabilize for at least one hour until the working temperature is reached before you start analysis.
Turning on the Lamp Automatically
To turn on the VIS lamp automatically after power on, in the chromatography data system, set AutoactivateVisible_Lamp to On.