The power supply of the device has wide-ranging capability, accepting any line voltage in the range specified for the device.
CAUTION
Electric Shock or Damage to the Device
Connecting the device to a line voltage higher or lower than specified could result in personal injury or damage to the device.
Connect the device to the specified line voltage only.