Flow cells and flow cell accessories
Item | Part No. |
---|---|
Standard flow cell, SST, 10 mm | 6077.0250 |
Semi-micro flow cell, SST, 7 mm | 6077.0360 |
Standard Bio flow cell, PEEK, 10 mm | 6077.0200 |
Semi-micro Bio flow cell, PEEK, 7 mm | 6077.0300 |
Nano flow cell, fused silica, 10 mm* | 6074.0270 |
Capillary flow cell, fused silica, 10 mm* | 6074.0280 |
UV monitor, fused silica, 45 nL | 6074.0285 |
Semi-preparative flow cell, PEEK, 0.4 mm | 6074.0320 |
Diagnostic cell | 6077.0190A |
Flushing and injection kit for flow cells, including syringe | 6078.4200 |
Viper capillary, 0.13 x 450 mm, stainless steel For connecting column and flow cell inlet when using UltiMate 3000 flow cells | 6040.2345 |
* There is no exact definition of the light path in capillary flow cells as is for conventional flow cells. Therefore, you should not use this value for calculating the absorption coefficient or the detector sensitivity according to the Lambert-Beer law.
TIP
UltiMate 3000 variable wavelength detector flow cells have the connection unit on the left side. To connect the column outlet to the connection unit installed on the left side of the flow cell in a Vanquish variable wavelength detector, Thermo Fisher Scientific recommends using the capillary of 450 mm length listed in the table above.
Miscellaneous
Item | Part No. |
---|---|
VIS lamp (tungsten lamp) | 6083.2000 |
DAC board Provides two analog outputs. Contact Thermo Fisher Scientific Technical Support for installation. | 6083.0900 |