The ship kit includes the items listed in the table. The kit content is subject to change and may vary from the information in this manual. Refer to the content list included in the kit for the most recent information about the kit content at the time when the device is shipped.
VC-D40 ship kit
Item | Quantity in shipment |
---|---|
Partition panel plug for guiding insulated capillaries | 1 |
System interlink cable | 1 |
Tubing bracket | 1 |
USB cable, USB 2.0 high-speed, type A to type B | 1 |
Viper capillary, I.D. x length 0.13 x 350 mm, stainless steel, connection column compartment – detector | 1 |
Waste line | 1 |
VF-D40 ship kit
Item | Quantity in shipment |
---|---|
Partition panel plug for guiding insulated capillaries | 1 |
System interlink cable | 1 |
Tubing bracket | 1 |
USB cable, USB 2.0 high-speed, type A to type B | 1 |
Viper capillary, I.D. x length 0.1 x 350 mm, MP35N, connection column compartment – detector | 1 |
Waste line | 1 |
For reordering information, see Consumables and Replacement Parts.