This section describes cleaning, replacement, storage and shipment of flow cells.
To store or ship the flow cell, follow these steps:
  1. Flush the flow cell with isopropanol.
  2. Remove the flow cell from the detector. See Removing the Flow Cell.
  3. Install the plugs on the flow cell inlet (IN) and outlet (OUT), and place the flow cell in its packaging.
To exchange the flow cell, follow these steps:
  1. Flush the flow cell with isopropanol.
  2. Remove the flow cell from the detector. See Removing the Flow Cell.
  3. Install a new flow cell without connection unit to the detector. See Installing a Flow Cell without Connection Unit.
    – or –
  4. Install a new flow cell with connection unit to the detector. See Installing a Flow Cell with Connection Unit.