The data collection rate is the number of data points per second (Hz) that the Chromeleon software collects from the detector and stores as raw data.
When you select a data collection rate, observe the following guidelines:
  • In general, each peak should be defined by at least 20 data points. For chromatograms with co-eluting peaks or low signal-to-noise ratios, 40 data points per peak are recommended.
  • If the data collection rate is too low, the start points, maxima, and end points of peaks will not be determined accurately.
  • If the data collection rate is too high, data files may need more disk space and post-run analyses may require more processing time.
  • If all peaks are relatively wide, select a lower data collection rate (for example, 1.0 Hz). This saves disk space and allows for a faster display of data in the Chromeleon software.
  • If any peaks of interest are less than a few seconds, select a higher data collection rate (10.0 Hz, for example).
Multiple wavelength applications
In multiple wavelength applications, baseline noise increases compared to single-channel data acquisition. This is especially the case if the data rate is set to the maximum or close to the maximum. To improve the signal-to-noise ratio, select a lower data collection rate.