The following table serves as an overview of parameters that influence the detector performance and indicates the performance characteristics affected.
Parameter
Affects
Wavelength
Sensitivity, linearity
Data collection rate
Peak resolution, disk space
Response Time/Peak width
Baseline noise, peak width, chromatographic resolution, sensitivity
Baseline behavior
Behavior of the baseline after switching the wavelength
For further information about the parameters, refer to the Chromeleon Help.