Parts required
Flow cell, as required by the application
Preparations
  1. Remove the diagnostic cell. See Removing the Diagnostic Cell.
  2. Unpack the flow cell.
Follow these steps
  1. Push the flow cell partly into the flow cell opening.
  2. Place the connection unit on the connection unit socket. Take care that the locating pin is placed in the slot.
  3. 4213
    Figure: Installing a flow cell with connection unit
    No.
    Description
    1
    Connection unit
    2
    Locating pin
    3
    Connection unit socket
    4
    Connection unit screw (knurl)
  4. Tighten the connection unit screw hand-tight.
  5. Push the flow cell completely into the flow cell opening.
  6. Tighten the two flow cell screws hand-tight.
  7. Set up the flow connections on the flow cell, see Setting Up the Flow Connections.
  8. Perform wavelength validation, see Performing a Wavelength Validation and Calibration.
TIP
If the wavelength validation fails, perform a calibration of wavelengths.