Parts required
Flow cell, as required by the application
Preparations
Remove the diagnostic cell. See Removing the Diagnostic Cell.
Unpack the flow cell.
Follow these steps
Push the flow cell into the flow cell opening.
Tighten the two flow cell screws hand-tight.
Set up the flow connections to the flow cell, see Setting Up the Flow Connections.
Perform wavelength validation, see Performing a Wavelength Validation and Calibration.
TIP
If the wavelength validation fails, perform a calibration of wavelengths.