The detector is shipped with a diagnostic cell installed. This section describes the removal of the diagnostic cell.
Follow these steps
  1. Open the doors.
  2. Loosen the two screws that hold the diagnostic cell in position.
  3. Grasp the diagnostic cell by its handle and pull the diagnostic cell out of the detector.
TIP
Store the flow cell in a clean place for later use in diagnostic procedures or for transporting the device.