This section describes the installation of the flow cell in the detector.
To remove the diagnostic cell, see Removing the Diagnostic Cell.
To install a flow cell without connection unit, see Installing a Flow Cell without Connection Unit.
To install a flow cell with connection unit, see Installing a Flow Cell with Connection Unit.
For instructions on cleaning or removing a flow cell or installing a diagnostic cell, see Flow Cell.
NOTICE
Sensitive Flow Cells
Flow cells are highly sensitive to dirt and dust. Observe the following notes when installing the flow cell to the detector:
NOTICE
Flow Cell Opening
The optical ports and the contact pad for the identification chip in the flow cell opening are sensitive to electrostatic discharge, contamination and scratches.