The user-accessible components of the device are located directly behind the front doors:
1234567891011
Figure: Interior components (here with flow cell with connection unit installed)
No.
Description
1
Cooling air intake
2
Keypad with status indicators
3
Screw holes for connection unit (left-side installation)
4
Flow cell with connection unit
5
Connection unit
6
Leak tray
7
Type label, indicating the module name, serial number, part number, and revision number (if any)
8
Drain port
9
Leak sensor
10
Partition panel
The recesses in the partition panel are used to route capillaries with the help of special plugs (see Installing the Partition Panel Plugs).
11
Lamp house cover
Depending on the detector configuration, a UV lamp or a UV and a VIS lamp are installed in the lamp house.