There are two types of partition panel plugs available in the detector ship kit.
Plugs available for the partition panel
Figure: Plugs available for the partition panel
No.
Description
1
Plug with slit, for guiding capillaries with small outer diameter, such as uninsulated capillaries.
2
Rotating plug, for guiding capillaries that do not fit in the plug with slit, including insulated capillaries.
Installing the plug with slit
  1. On the detector partition panel, push the plug with slit in the required recess of the partition panel (if not present yet).
  2. To secure the plug with slit, insert the nose into the opening of the partition panel.
  3. Securing the partition panel plug with slit
    Figure: Securing the partition panel plug with slit
  4. To secure the capillary, push the capillary into the slit of the plug.
Capillary installed in the plug with slit
Figure: Capillary installed in the plug with slit
Installing the rotating plug
  1. On the detector partition panel, push the rotating plug in the required recess of the partition panel (if not present yet).
  2. To open the plug in order to route the capillary through the plug, turn the rotating plug toward the front.
  3. To secure the capillary, turn the rotating plug toward the partition panel to close the opening of the rotating plug.
Using the rotating plug
Figure: Using the rotating plug