Pay attention to the following safety guidelines:
Observe all warning messages and precautionary statements presented in Safety Precautions.
CAUTION
Heavy Load, Bulky Device
The device is too heavy or bulky for one person alone to handle safely. To avoid personal injury or damage to the device, observe the following guidelines:
  • Physical handling of the device, including lifting or moving, requires a team effort of two persons.
  • A team effort is in particular required when lifting the device into the system stack or when removing it.
  • Use the carrying handles that were shipped with the device to move or transport the device. Never move or lift the device by the front doors. This will damage the doors or the device.
CAUTION
Electric Shock or Damage to the Device
After the power to the device is turned off, the device is still energized as long as the power cord is connected. Repair work on the device while the device is connected to power could lead to personal injury.
  • Always unplug the power cord before starting repair work inside the device.
  • If you were instructed to remove any housing covers or panels, do not connect the power cord to the device while the cover or panels are removed.