The detector design allows easy access to the flow cell on the interior front.
1124635
Figure: Flow cell (example)
No.
Description
1
Flow cell screws
2
Flow cell handle
3
Drain outlet
4
Connector for outlet capillary
5
Connection unit screw
6
Connector for inlet capillary
Flow Cell Label
One or more flow cell labels are present on the flow cell, which contain information such as flow cell type, part number, and serial number.
Flow Cell Identification Chip
An identification (ID) chip on the flow cell stores information, including the flow cell type and the serial number of the flow cell. The ID chip also stores data during operation, such as the exposure time to the light.
When the flow cell is installed, the detector reads the data from the chip and transfers the flow cell data to the chromatography data system.
Heat exchanger
Standard flow cells and semi-micro flow cells are equipped with a built-in heat exchanger.
The heat exchanger helps to adapt the temperature of the mobile phase to the flow cell temperature before the mobile phase enters the optical flow path within the flow cell. Note that the volume of the heat exchanger and inlet capillary influences the retention times and peak widths.
Flow Cell Types
The detector is shipped with a diagnostic cell. Replace the diagnostic cell with a flow cell that is appropriate for your application. Depending on the application needs, different types of flow cells are available for the detector.
For flow cell ordering information, see Optional Accessories.
For the flow cell specifications, including, for example, materials in the flow path and pressure limit, see Flow Cell Specifications.
For details about the flow cells or about the availability of other flow cells, refer to the Thermo Fisher Scientific sales organization.