To prepare the device for transport, follow these steps:
  1. Perform a long-term shut down of the device (see Long-Term Shutdown).
  2. Turn off the device with its main power switch and disconnect the power cord.
  3. Remove all cables and flow connections to other devices.
  1. Remove the flow cell from the detector, and store the flow cell in its packaging. Install the diagnostic cell to the flow cell opening of the detector.
NOTICE
The flow cell opening on the device is sensitive to dust and debris.
If no flow cell is installed to the detector, install a diagnostic cell to the flow cell opening of the detector.
  1. Remove the device or slide-in module from the system stack as required:
    Install the carrying handles and remove the device from the Vanquish system. Follow the instructions on dismounting the system stack in the Transporting or Shipping the System section of the Vanquish System Operating Manual.
    ‑or‑
    Remove the slide-in module from the device enclosure in the system stack (see Removing the Slide-In Module).