NOTICE
Flow cells are highly sensitive to dirt and dust. Observe the following notes when handling flow cells:
  • When holding flow cells, do not touch the optical block of the flow cell or the sensitive electronics on the flow cell rear side.
  • The optical ports of the flow cell are sensitive to contamination and scratches. Do not touch the optical ports of the flow cell or immerse them.
  • To avoid damage to the optical ports of the flow cell, be careful when inserting the flow cell into the flow cell opening of the device.
  • On the rear side of the flow cell, the contact pads for the identification chip are located. Never touch the contact pads. Avoid damage to the electronics of the ID chip.
  • Particulate matter, dust and debris can lead to contamination and clogging of the flow cell. If capillaries are removed from the flow cell inlet and outlet, install protective plugs to close the ports and to prevent particles from clogging the flow cell in the next application. Always use the plugs that were installed when the flow cell was shipped.
  • Use the dedicated packaging when storing or transporting the flow cell.