If the device will not be operated for some time, follow the instructions in this section to shut down the device.
TIP
The Chromeleon software provides procedures for automatically preparing the chromatography system for shutdown. The procedures include, for example, operations for reducing the flow rate, reducing the temperature in temperature-controlled devices, and turning off the detector lamps.
For information about Smart Shutdown and Smart Standby, refer to the Chromeleon Help.