The inlet and outlet ports of the flow cells support the following capillary fittings:
  • Viper™
  • nanoViper™
  • Standard 1/16" HPLC fittings using ferrules (PEEK, stainless steel) or finger-tight fittings (PEEK)
This section provides information about how to connect and handle capillaries, fittings, and tubing.