Preparations
  1. See Flow Connections to the Flow Cell.
Follow these steps
123
Figure: Connecting the detector waste line
  1. Connect the waste line to the flow cell outlet or to the outlet (OUT) on the flow cell connection unit (1).
  2. Route the waste line through the top recess (2) in the partition panel.
  3. Route the waste line through the tubing guides of the system modules below the detector to the Vanquish system base (3).
  4. On the system base, route the detector waste line through the dedicated detector waste outlet to waste and connect the detector waste line to the waste container as described in the Vanquish System Operating Manual.
TIP
The waste line should go straight to the system base and to waste. Make sure that the line is positioned straight in the tubing guides.
  1. If you have to cut tubing to length, use a tubing cutter. Make sure that the cut is at right angle to the length of the line and that the cut is not crimped.
  2. Check the waste line over the entire flow path: Make sure that no bending (kink), pinching or squeezing of the waste line is present at any point in the flow path.