The table lists the safety symbols that appear on the device or on labels affixed to the device. Follow the safety notices in this manual to prevent the risk of operator injury or damage to the device.
Symbol | Description |
---|---|
Indicates a potential hazard. Refer to this manual to avoid the risk of personal injury and/or to prevent damage to the device. | |
— Ο | Power supply is on Power supply is off |
Indicates alternating current. | |
Indicates that the surface becomes hot during operation. Do not touch these surfaces while they are heated up. | |
Indicates that the UV radiation produced by the deuterium lamp in the device may be harmful to eyes and skin. Do not look directly into the light produced by the deuterium lamp. Never operate the lamp outside the device. |