Manual purging requires user interaction. The user must open and close the purge valve by hand to direct the solvent to waste, through the purge outlet on the purge/sensor unit.
The procedure depends on the reason for purging: