Solvent shut-off valve
Description | Part No. |
---|---|
Solvent shut-off valve To prevent the solvent from flowing through the system, for example, when opening a flow connection on the low-pressure side. | 6036.0010 |
Solvent monitor
Description | Part No. |
---|---|
Vanquish Solvent Monitor To actively monitor the liquid level in solvent reservoirs and waste containers in real time. Monitoring is based on real physical measurement. | |
Solvent monitor, 4-channel version | 6230.1320 |
Solvent monitor, 8-channel version | 6230.1310 |
Buffer Performance kit
Description | Part No. |
---|---|
Buffer Performance kit for quaternary or dual pumps To avoid pressure pulsation issues with buffer applications in quaternary pumps or dual pumps. Depending on the application, two kits may be required with the dual pump. Due to bad solubility of salts in organic fluids, mixing of a buffer solution with organic fluids in a quaternary or dual pump can lead to precipitation of salt at the mixing point. The precipitated salt can deposit in the check valves and prevent them from closing. This leads to pressure pulsation in buffer applications. Modifying the sequence of the fluidics with the kit components enhances the mixing performance. | 6040.5320 |
Degasser kit (VC-P40)
Description | Part No. |
---|---|
Degasser kit VC-P40 To retrofit the isocratic pump with a vacuum degasser. Thermo Fisher Scientific service personnel must retrofit the pump with the degasser. | 6045.3731 |
Mixing systems, inline filters
Description | Part No. |
---|---|
Mixing systems, set inline filter |
|