Each mixing system comprises a static mixer and a capillary mixer. The volumes of both mixers determine the entire volume of the mixing system.
TIP
With mixing systems (volumes: 200 µL, 400 µL, 800 µL or 1550 µL), only the static mixer is different from the static mixer with which the pump is shipped. The capillary mixer remains the same. Thus, if you want to change to this mixing system, you need to order only the static mixer. If you want to replace the capillary mixer nevertheless, order the appropriate part number as indicated in the table.
Static mixers
Description
Part No.
Static mixers for use with 50 µL capillary mixer:
Static mixer, volume: 150 µL
(for total volume of mixing system: 200 µL)
Binary VF-pump, isocratic pump: The pump is shipped with the mixing system with a volume of 200 µL.
With the isocratic pump, the mixing system serves as inline filter.
6044.5110
Static mixer, volume: 350 µL
(for total volume of mixing system: 400 µL)
Binary VC-pump, quaternary pump: The pump is shipped with the mixing system with a volume of 400 µL.
Dual pump: The pump is shipped with two mixing systems, each with a volume of 400 µL.
6044.5310
Static mixer, volume: 750 µL
(for total volume of mixing system: 800 µL)
6044.5750A
Static mixer, volume: 1500 µL
(for total volume of mixing system: 1550 µL)
6044.5450A
Capillary mixers
Description
Part No.
Capillary mixer (volume: 50 µL) for use with static mixers
(volumes: 150 µL, 350 µL, 750 µL, and 1500 µL) for:
VC-pumps
6044.3015
VF-pumps
6044.5026
Mixing systems, set inline filter
VC-pumps
Description
Part No.
Mixing system, volume: 100 µL, including:
  • Static mixer, volume: 75 µL
  • Capillary mixer, volume: 25 µL
6045.5100
Set inline filter, volume: 35 µL, including:
  • Inline filter (static filter) volume: 10 µL
  • Capillary mixer, volume: 25 µL
In addition, order the capillary to connect the pump to the autosampler.
6045.3020
Capillary to connect the pump to the autosampler, for use with 100 µL mixing system and 35 µL inline filter set
For use with binary pump, isocratic pump, and quaternary pump
Dual pump: The capillaries to use depend on the application workflow and are provided in the respective Workflow Solution Kits. For details refer to the documentation that comes with the kit.
6040.2325
VF-pumps
Description
Part No.
Mixing system, volume: 100 µL, including:
  • Static mixer, volume: 75 µL
  • Capillary mixer, volume: 25 µL
6044.5100
Set inline filter, volume: 35 µL, including:
  • Inline filter (static filter) volume: 10 µL
  • Capillary mixer, volume: 25 µL
In addition, order the capillary to connect the pump to the autosampler.
6044.3870
Capillary to connect the pump to the autosampler, for use with the 100 µL mixing system and 35 µL inline filter set
For use with binary pump, isocratic pump, and quaternary pump
Dual pump: The capillaries to use depend on the application workflow and are provided in the respective Workflow Solution Kits. For details refer to the documentation that comes with the kit.
6042.2330