TIP
Maintenance kits can include parts that are not required for your specific pump. These parts are marked accordingly in the contents lists for the kits.
VC-P10
Description
Part No.
Maintenance kit for VC-P10, including:
  • Pump head seal (2 PTFE seals (2 each), different sized)
  • Silicone tubing (clear tubing)
  • Peristaltic tubing (light yellow tubing)
  • Tubing connector (straight) (pack of 5)
  • Tubing connector (90°-angled)
  • Seal wash seal (pack of 4)
  • Piston seal (pack of 4)
  • Valve cartridge, ceramics (pack of 2)
  • Solvent line filter (4 filter holders and filter frits)
  • Cleaning swab (pack of 5)
  • Tubing clip (self-adhesive) (pack of 3)
Not required with VC-P10: Support ring
6040.1953A
VC-P20
Description
Part No.
Maintenance kit for VC-P20, including:
  • Pump head seal (2 PTFE seals, different sized)
  • Silicone tubing (clear tubing)
  • Peristaltic tubing (light yellow tubing)
  • Tubing connector (straight) (pack of 4)
  • Tubing connector (90°-angled)
  • Seal wash seal (pack of 2)
  • Piston seal (pack of 2)
  • Valve cartridge, ceramics
  • Solvent line filter (4 filter holders and filter frits)
  • Cleaning swab (pack of 5)
  • Tubing clip (self-adhesive) (pack of 2)
Not required with VC-P20: Support ring
6040.1951A
VC-P32
Description
Part No.
Maintenance kit for VC-P32, including:
  • Pump head seal (2 PTFE seals (2 each), different sized)
  • Silicone tubing (clear tubing)
  • Peristaltic tubing (light yellow tubing)
  • Tubing connector (straight) (pack of 5)
  • Tubing connector (90°-angled)
  • Seal wash seal (pack of 4)
  • Piston seal (pack of 4)
  • Valve cartridge, ceramics (pack of 2)
  • Solvent line filter (6 filter holders and filter frits)
  • Cleaning swab (pack of 5)
  • Tubing clip (self-adhesive) (pack of 3)
Not required with VC-P32: Support ring
6040.1952A
VC-P40
Description
Part No.
Maintenance kit for VC-P40, including:
  • Pump head seal (2 PTFE seals, different sized)
  • Silicone tubing (clear tubing)
  • Peristaltic tubing (light yellow tubing)
  • Tubing connector (straight) (pack of 3)
  • Tubing connector (90°-angled)
  • Seal wash seal (pack of 2)
  • Piston seal (pack of 2)
  • Valve cartridge, ceramics
  • Solvent line filter (1 filter holder and filter frits)
  • Cleaning swab (pack of 5)
  • Tubing clip (self-adhesive) (pack of 2)
Not required with VC-P40: Support ring
6040.1950A