For reordering information of the ship kit components, see Consumables and Replacement Parts.
VC-P32, VC-P33 ship kit
Item
Quantity in shipment
Silicone tubing, 3 m
1
Syringe, plastic, 12 mL
1
Tubing connector (90°-angled)
2
Solvent line filter, filter holder (no filter frit included)
6
Solvent line filter, filter frit, 10 µm (pack of 10)
1
Plugs and retaining guides for reservoir caps, kit including
  • Cap plug to close open holes in the reservoir cap (pack of 5)
  • Retaining guide to keep the liquid line in place in the reservoir cap (pack of 2)
7
Fitting plug, Viper
1
Tool kit, including
  • Open-end wrench, size 11 x 13 mm
  • Double-ended ring wrench, size 11 x 13 mm
  • Open-end wrench, size 1/4" x 5/16"
  • Hexagon wrench, size 6
  • Seal-handling tool
  • Spacing tool, pump head and pistons
1
Reservoir, 0.25 L, with reservoir cap
1
Tubing bracket
1
Solvent lines (pack of 6) from solvent reservoirs to pump degasser
1
System interlink cable (RJ45), 0.5 m
1
USB cable, type A to type B, high-speed, USB 2.0, cable length: 5 m
1
VF-P32 ship kit
Item
Quantity in shipment
Silicone tubing, 3 m
1
Syringe, plastic, 12 mL
1
Tubing connector (90°-angled)
2
Solvent line filter, filter holder (no filter frit included)
6
Solvent line filter, filter frit, biocompatible, 10 µm
6
Plugs and retaining guides for reservoir caps, kit including
  • Cap plug to close open holes in the reservoir cap (pack of 5)
  • Retaining guide to keep the liquid line in place in the reservoir cap (pack of 2)
7
Fitting plug, Viper
1
Tool kit, including
  • Open-end wrench, size 11 x 13 mm
  • Double-ended ring wrench, size 11 x 13 mm
  • Open-end wrench, size 1/4" x 5/16"
  • Hexagon wrench, size 6
  • Seal-handling tool
  • Spacing tool, pump head and pistons
1
Installation tool for Viper capillaries with torque toothing
1
Reservoir, 0.25 L, with reservoir cap
1
Tubing bracket
1
Solvent lines (pack of 6) from solvent reservoirs to pump degasser
1
System interlink cable (RJ45), 0.5 m
1
USB cable, type A to type B, high-speed, USB 2.0, cable length: 5 m
1