The picture illustrates how the pump operates:
Operating principle
Figure: Operating principle
No.
Description
A
Solvent
1
Pump head, with
1a
Working cylinder
1b
Equilibration cylinder
2
Purge/sensor unit, with
2a
Purge valve knob
2b
Purge outlet
2c
System pressure sensor
3
Mixing system, serving as inline filter, with
3a
Capillary mixer
3b
Static mixer
4
Pump outlet (outlet of the static mixer)
For additional information, see Operating Principle.