The user-accessible components of the pump are located directly behind the front doors:
Interior view
Figure: Interior view
No.
Description
1
Keypad with buttons for pump control
2
Pump head
3
Purge/sensor unit including purge valve and system pressure sensor
4
Leak sensor
5
Type label, indicating the module name, serial number, part number, and revision number (if any)
6
Drain port for guiding leaking liquid to waste
7
Seal wash detector (drop detector)
8
Seal wash pump
9
Tubing chase with tubing guides
10
Pump block LED
11a
Capillary mixer (as part of mixing system, which serves as inline filter)
11b
Static mixer (as part of mixing system, which serves as inline filter)
12
Pump outlet (outlet of the static mixer)