If you are unable to remove air bubbles with purging, purge the pump against backpressure. Follow the procedure for running in new piston seals in Recommendations for New Piston Seals.
If you are unable to remove air bubbles with purging, purge the pump against backpressure. Follow the procedure for running in new piston seals in Recommendations for New Piston Seals.