These steps apply only when installing a shut-off valve during initial setup of the pump. To install a shut-off valve after initial operation of the pump, follow the steps in Installing Solvent Shut-Off Valves.
When
To shut off the solvent flow through the system at the solvent reservoir, for example, to prevent solvent from flowing through the system when you open a flow connection on the low-pressure side
Parts and additional items required
  • Shut-off valve, kit containing one shut-off valve and fitting connections
  • Tubing cutter
Preparations
Prepare the pump and follow the steps in Connecting the Solvent Lines.
Follow these steps
Installing a shut-off valve in the solvent line
Figure: Installing a shut-off valve in the solvent line
No.
Description
1
Solvent line
2
Fitting
3
Ferrule
4
Shut-off valve
  1. Cut the solvent line about 10 cm from the reservoir cap with a tubing cutter. Make sure that the cut is at a right angle to the length of the line.
  2. Onto the free end of each solvent line, slide a fitting and a ferrule. Mind the correct orientation of the fitting and ferrule (see picture).
  3. Tighten the solvent lines to the shut-off valve.
Solvent line with shut-off valve installed
Figure: Solvent line with shut-off valve installed
No.
Description
1
Shut-off valve
2
Retaining guide
  1. Check the solvent lines across the entire flow path. Make sure that the lines are not bent, pinched, or squeezed at any point in the flow path.