This section applies to Vanquish Core system modules.
In a Vanquish Core system, normal-phase (NP) compatible solvents and additives may be used if the system modules have been modified for NP applications. Refer to the Vanquish System Operating Manual.