Reservoirs for solvents and wash liquids
Description | Part No. |
---|---|
Reservoir, 1 L, including cap | 2270.0012 |
Reservoir, 0.25 L, including cap | 2270.0026 |
Cap for reservoirs, screw-cap (pack of 4) | 6270.0013 |
Cap plug to close open holes in the reservoir cap (pack of 20) | 6000.0047 |
Retaining guide to keep the liquid line in place in the reservoir cap (pack of 5) | 6000.0042 |
Plugs and retaining guides for reservoir caps, kit including
| 6030.9101 |
Seal wash system
Description | Part No. |
---|---|
Peristaltic and wash tubing kit The kit includes peristaltic tubing (PharMed), silicone tubing, and tubing connectors for use in
NOTICE: Use the thick silicone tubing (and related tubing connectors) in the autosampler. Use the thin silicone tubing (and related tubing connectors) in the pump. Use the screwable connectors for the pump head. The screwable tubing connectors are not used with the VC‑pumps and VF‑pumps. NOTE: For a Vanquish Core system that has been modified for NP applications, order the Normal-Phase (NP) tubing kit. For ordering details and kit content, refer to the Vanquish System Operating Manual. | 6044.1150 |
Seal wash detector (drop detector) with funnel | 6044.1898 |
Seal wash detector (drop detector) with funnel, NP | 6044.1902 |
Seal wash seals | |
VF‑pumps: Seal wash seal (pack of 2 seal wash seals and 2 piston seals (RP)) | 6044.0295 |
VC‑pumps – all VC‑pumps except VC‑P21 and VC‑P33: Seal wash seal (pack of 2 seal wash seals) | 6040.0033 |
VC‑P21, VC‑P33, and VC-pumps modified for NP applications: Seals (NP) (pack of 2) for use as both seal wash seals and piston seals | 6040.0306 |
Solvent line filters, solvent lines and tubing
Description | Part No. |
---|---|
Solvent line filter, filter holder (pack of 6) (no filter frits included) | 6268.0115 |
Solvent line filter, filter frits, 10 µm (pack of 10) for | |
VF‑pumps (biocompatible frits) | 6268.0111 |
VC‑pumps (stainless steel frits) | 6268.0110 |
Solvent lines to connect the solvent reservoirs to the pump inlet | |
Quaternary pump: Solvent lines to connect the solvent reservoirs to the degasser inlet ports | 6036.1703 |
Dual pump: Solvent lines to connect the solvent reservoirs to the degasser inlet ports | 6036.1704 |
Binary pump: Solvent lines to connect the solvent reservoirs to the solvent selectors | 6036.1701 |
Isocratic pump: Solvent line to connect the solvent reservoir to the pump head | 6036.1705 |
Solvent lines to connect the pump inlet to the pump heads | |
Quaternary pump: Tubing kit, including:
| 6044.8017 |
Dual pump: Tubing kit, including:
| 6044.8005 |
Binary pump: Tubing kit, including:
| 6044.8006 |