Each mixing system comprises a static mixer and a capillary mixer. The volumes of both mixers determine the entire volume of the mixing system.
The tables list the reordering part numbers for the components with which the pump is shipped from the factory. Mixing systems with other volumes are available as an option. For ordering information, see Optional Accessories.
Static mixers
Description | Part No. |
---|---|
Static mixer, volume: 350 µL | 6044.5310 |
Static mixer, volume: 150 µL | 6044.5110 |
Capillary mixers
Description | Part No. |
---|---|
Capillary mixer (volume: 50 µL) for use with static mixers | |
VC-pumps | 6044.3015 |
VF-pumps | 6044.5026 |