Parts required
  • Set inline filter, volume: 35 µL, comprising an inline filter (static filter, volume: 10 µL) and a capillary mixer (volume: 25 µL)
  • Capillary to connect the pump to the autosampler
Tools and additional items required
  • Depending on the fitting connection on the capillaries
    • Viper capillary: Viper knurl (usually present on the capillary)
    • Conventional capillary (non-Viper): Open-end wrench, size 1/4"
  • Solvent suitable for purging
  • Screwdriver, Torx T10
Preparations
  1. If not yet done: Remove the mixing system with which the pump is shipped (see Removing the Mixing System).
  1. The inline filer requires a different mounting bracket. Install the new mounting bracket. The mounting bracket is included in the inline filter ship kit.
Installing the New Mounting Bracket
  1. Unplug the system pressure sensor cable from the P-SYS connector.
  2. Remove the seal wash detector assembly (funnel and detector). Grasp and pull the assembly upward by the funnel. The detector including the tubing and cable can remain connected to the funnel.
Removing the seal wash detector assembly
Figure: Removing the seal wash detector assembly
No.
Description
1
Funnel, seal wash detector
2
Seal wash detector
3
Detector cable
4
Seal wash line
  1. To attach the new mounting bracket inside the pump (above the purge/sensor unit):
  1. Remove the push rivets that cover the mounting holes while no mounting bracket is installed.
  2. Using the screws shipped with the mounting bracket, attach the new bracket inside the pump.
Installing the mounting bracket
Figure: Installing the mounting bracket
No.
Description
1
Mounting holes for mounting bracket (covered by push rivets while no mounting bracket is installed in this position)
2
Mounting bracket
3
Mounting screws (Torx)
Follow these steps
  1. Connect the capillary mixer on the purge/sensor unit. Mind the direction of flow through the capillary mixer (indicated by the arrow on the mixer).
  2. TIP
    To facilitate installation of the inline filter, loosely tighten the capillary mixer on the purge/sensor unit so that you can easily access the free end of the capillary.
  3. Connect the inline filter to the capillary mixer. Mind the direction of flow through the inline filter (indicated by the arrow on the filter).
  4. Turn and insert the assembly of inline filter and capillary mixer into the mounting bracket.
Inline filter with capillary mixer installed
Figure: Inline filter with capillary mixer installed
No.
Description
1
Inline filter
2
Capillary mixer
  1. Tighten the capillary mixer on the purge/sensor unit.
  2. Reinstall the seal wash detector assembly in the pump (see Replacing the Seal Wash Detector).
  3. Reconnect the system pressure sensor cable.
  4. Connect the inline filter outlet to the autosampler injection valve (see Connecting the Pump and Autosampler).
  5. In the Chromeleon software, set the StaticMixer property to the value indicated on the inline filter. If the property is not set to the correct value, the leak tests may not provide reliable results.
  6. With Chromeleon 7.2.8 and later: In the Chromeleon software, set the CapillaryMixer property to the value indicated on the capillary mixer.
  7. Apply the flow rate of your application and have the pump deliver for a short time.
  8. Inspect the flow connections on the mixer for indications of leakage before resuming operation.
  9. In the Chromeleon software, consider updating the Predictive Performance information for the static mixer (MixerChanged command).
When operating the pump, observe the following
  • Operating the pump with the inline filter reduces the gradient delay volume but increases the ripple.
  • The maintenance procedures for the static mixer apply equally to the inline filter (see Mixing System).