Every six weeks, the pump automatically performs an internal maintenance procedure when you initiate a purge cycle. The purge cycle starts automatically when internal maintenance is complete. This may take some seconds. While internal maintenance is running, the purge valve must remain open.
If no purge cycle is initiated after another 6 weeks, a message appears in the Instrument Audit Trail, reminding you to start a purge cycle.